Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

93295

Sigma-Aldrich

TRIS acetate – EDTA buffer substance

BioReagent, 10× concentrate, powder, white

Synonym(s):

TAE buffer dry blend

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105319
NACRES:
NA.25

grade

Molecular Biology

product line

BioReagent

form

powder

quality

10× concentrate

impurities

DNases, none detected
RNases, none detected
phosphatases, none detected
proteases, none detected

color

white

pH

8.2-8.5

solubility

water: 64.2 g/L, clear, colorless

Reconstitution

dissolution to the indicated volume yields a 10× concentrated solution (0.4 M TRIS acetate, pH ~8.5, 10 mM EDTA.Na2)

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service