Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

93220-001

Sigma-Aldrich

RNAstable®

Trial Kit, for room temp preservation of DNA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

grade

Molecular Biology
for molecular biology

form

dry

usage

3 on-column digestions

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

storage temp.

room temp

General description

SampleMatrix® by Biomatrica® is a proprietary core technology to stabilize and protect biological materials at room temperature without degradation. The SampleMatrix technology was designed by combining extremophile biology that enables long-term survival of organisms in extremely dry environments, with synthetic chemistry.

RNAstable® preserves RNA samples at room temperature. Each tube or plate contains RNAstable® as a coating at the bottom of the tube or well, which protects picogram to microgram amounts of RNA. This medium is completely dissolvable and ensures total and quantitative sample recovery.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product was Designed for Energy Efficiency. To see how Stanford incorporated Biomatrica Technology and saved substantial amounts of energy, see their published study:

Biomatrica technology enables safe storage of biological material at room temperature.

Features and Benefits

  • Preserves RNA at ambient temperatures without degradation
  • Eliminates the need to store RNA in freezers or to ship on dry ice
  • Concentrates RNA more effectively than other methods
  • Compatible with most downstream applications

Packaging

  • (3) RNAstable® Tubes
  • 1.5 mL screw-cap microfuge tube made of durable polypropylene
  • (1) tube of sterile water
  • Quick Reference Protocol
  • Barcode-labeled for sample tracking
  • Enclosed in re-sealable moisture-barrier foil pouch

Legal Information

Biomatrica is a registered trademark of Biomatrica
RNAstable is a registered trademark of Biomatrica
SampleMatrix is a registered trademark of Biomatrica

related product

Product No.
Description
Pricing

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wen Huang et al.
PloS one, 7(1), e30244-e30244 (2012-02-01)
In this study, mRNA-Seq was used to characterize and compare the leukocyte transcriptomes from two taurine breeds (Holstein and Jersey), and one indicine breed (Cholistani). At the genomic level, we identified breed-specific base changes in protein coding regions. Among 7,793,425
Gilberto E Hernandez et al.
BioTechniques, 47(2), 667-668 (2009-09-10)
RNA integrity is a critical factor in obtaining meaningful gene expression data. Current methodologies rely on maintaining samples in cold environments during collection, transport, processing, and storage procedures, which are also extremely time-sensitive. Several RNA storage products are commercially available
Stephen K Gire et al.
Science (New York, N.Y.), 345(6202), 1369-1372 (2014-09-13)
In its largest outbreak, Ebola virus disease is spreading through Guinea, Liberia, Sierra Leone, and Nigeria. We sequenced 99 Ebola virus genomes from 78 patients in Sierra Leone to ~2000× coverage. We observed a rapid accumulation of interhost and intrahost
Daniel S Stevens et al.
Journal of virological methods, 182(1-2), 104-110 (2012-03-22)
Two approaches to stabilize viral nucleic acid in processed clinical specimens were evaluated. HIV-1 RNA extracted from clinical specimens was stabilized in a dry matrix in a commercial product (RNAstable, Biomatrica, San Diego, CA, USA) and in a reverse-transcription reaction
Adrian Puren et al.
The Journal of infectious diseases, 201 Suppl 1, S27-S36 (2010-03-17)
RNA remains the most informative and accurate biomarker for human immunodeficiency virus type 1 load diagnostics and for surveillance of drug resistance markers. Viral load testing by nucleic acid amplification currently is a complex and expensive test that is restricted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service