Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

93020-017

Sigma-Aldrich

DNAstable® Blood

Trial Kit, for room temp preservation of DNA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

grade

Molecular Biology

form

dry

usage

3 reactions

storage temp.

room temp

General description

Stabilize and Preserve DNA in Whole Blood at Room Temperature. Preserve DNA in blood and buffy coat samples for at least 12 years at ambient temperatures. Cost effective and convenient blood sample archiving. Compatible with all downstream applications.

Packaging

1 x 3 kit

Physical form

  • 3 Tubes DNAstable Blood
  • Screw-cap tube made of durable polypropylene
  • Barcode labeled for sample tracking
  • Quick Reference Protocol
  • Enclosed in re-sealable moisture-barrier foil pouch

Legal Information

DNAstable is a registered trademark of Biomatrica

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Malini Udtha et al.
Biopreservation and biobanking, 12(5), 332-336 (2014-10-24)
Preservation of biospecimens for biobanking applications traditionally involves freezing while maintaining the integrity of the product throughout multiple freeze-thaw cycles. The protection and stabilization of DNA at room temperature (RT) may eliminate the costs associated with freezer storage and reduce

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service