Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

92984

Sigma-Aldrich

Streptavidin B-Phycoerythrin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

form

liquid

mol wt

Mr ~240000

fluorescence

λex 542 nm; λem 575 nm in 0.1 M phosphate pH 7.0

suitability

corresponds for gel electrophoresis

storage temp.

2-8°C

Physical form

1 mg/ml in 0.1 M sodium phosphate, 0.1 M sodium chloride, 2 mM sodium azide, pH 7.5

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service