Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

92821

Sigma-Aldrich

Fluorescence Marker Kit 650

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

storage temp.

2-8°C

General description

This kit provides the required reagents for labeling proteins with Fluorescent Red 646. This label shows similar properties to Cy®5, such as strong fluorescence, spectral match with corresponding laser line, and good water solubility. Together with Fluorescent Orange 548 it is ideally suited for energy transfer studies, hybridization assays and flow cytometry.

Application

For fluorescent labeling of proteins, peptides, and amino-modified nucleotides. Comprises phosphate buffer solution, bicarbonate buffer solution, and Fluorescent Orange 646 reactive.

Quantity

Sufficient for 5 labeling reactions of 1.5 mg protein.

Legal Information

Cy is a registered trademark of Cytiva

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service