92592
Fluorescent orange 550 reactive
suitable for fluorescence, ≥90.0% (coupling to amines)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C36H47N4NaO11S
Molecular Weight:
766.83
MDL number:
UNSPSC Code:
12352200
assay
≥90.0% (coupling to amines)
form
powder
solubility
DMF: soluble
fluorescence
λex 550 nm; λem 590 nm in 0.1 M phosphate pH 7.0
suitability
suitable for fluorescence
storage temp.
−20°C
General description
Fluorescent orange 550 reactive is a fluorescent label especially well suited for excitation by He-Ne lasers (544 nm) or mercury arc lamps. Fluorescent orange 550 reactive shows strong fluorescence and may be used as alternative cyanine dyes.
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service