Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

92465

Sigma-Aldrich

Sodium citrate tribasic 0.2M – HEPES sodium salt pH 7.5; 0.1M – 2-propanol 20% solution

Synonym(s):

Crystallization solution 27/Kit-No 82009

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

form

liquid

pH

7.5±0.2 ( in neat)

density

1.01 g/mL at 20 °C

storage temp.

2-8°C

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alessandra Giorgetti et al.
Experimental hematology, 45, 85-93 (2016-10-25)
Recent studies in zebrafish and mice have revealed that proinflammatory signaling is a positive regulator of definitive hematopoietic development. Whether proinflammatory signaling also regulates human hematopoietic specification remains unknown. Here, we explored the impact of the proinflammatory cytokines tumor necrosis

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service