Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

92432

Sigma-Aldrich

Cellulose acetate sheets

unperforated, size × layer thickness 25 mm × 150 mm × 100-135 μm

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200

form

sheet

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

With cellulose acetate electrophoresis, you can separate nearly all sample materials that migrate in an electrical field. Apply 0.06 to 0.15 mg of sample mixture per cm application line. Concentrate the mixture to at least 1 mg/ml. Advantages of those strips include:
  • High porosity of the strip (approx. 80 % free pore volume)
  • Isotropic, integral matrix for ideal flow conditions
  • Minimal diffusion effect and low adsorption capacity
  • Perfect transparency because of the pore system
  • Ideal for the non-toxic ATX reagent system
  • Virtually unlimited stability at room temperature
  • Two identical sides, no need to search for the "right" side
Available in different sizes for all current chamber types

These strips can also be used for high-voltage electrophoresis, chromatography, and immunodiffusion.

Storage Class

13 - Non Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service