Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

92364

Sigma-Aldrich

Tetramethylrhodamine-6-maleimide

suitable for fluorescence, ≥50% (HPLC), ≥97% (isomer purity, HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H23N3O5
CAS Number:
Molecular Weight:
481.50
MDL number:
UNSPSC Code:
12352200

assay

≥97% (isomer purity, HPLC)

form

powder

concentration

≥50% (HPLC)

solubility

DMSO: soluble
methanol: soluble

fluorescence

λex 543 nm; λem 575 nm in 0.1 M phosphate pH 7.5

suitability

suitable for fluorescence

storage temp.

−20°C

InChI

1S/C28H23N3O5/c1-29(2)16-5-9-20-23(14-16)36-24-15-17(30(3)4)6-10-21(24)27(20)19-8-7-18(13-22(19)28(34)35)31-25(32)11-12-26(31)33/h5-15H,1-4H3

InChI key

GXCPCIODYFPPQV-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service