Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

92315

Sigma-Aldrich

Fluorescent red NIR 730 reactive

suitable for fluorescence, ≥90.0% (coupling to amines)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H51N3O8S
Molecular Weight:
757.93
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32

assay

≥90.0% (coupling to amines)

form

powder

solubility

DMF: soluble

fluorescence

λex 680 nm; λem ~755 nm in 0.1 M phosphate pH 7.0

suitability

suitable for fluorescence

storage temp.

−20°C

Analysis Note

further Exc.: 730 nm (results in a lower emission)

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service