Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

92173

Sigma-Aldrich

2,2′-Dimethoxybiphenyl

BioReagent, suitable for fluorescence, ≥99.0% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H14O2
CAS Number:
Molecular Weight:
214.26
Beilstein/REAXYS Number:
2051625
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

description

fluorescent marker

product line

BioReagent

assay

≥99.0% (HPLC)

form

crystals

fluorescence

λex 280 nm; λem 339 nm in ethanol (1 mg/mL)

suitability

suitable for fluorescence

storage temp.

2-8°C

SMILES string

COc1ccccc1-c2ccccc2OC

InChI

1S/C14H14O2/c1-15-13-9-5-3-7-11(13)12-8-4-6-10-14(12)16-2/h3-10H,1-2H3

InChI key

VGMKUVCDINAAFC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service