Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

92102

Sigma-Aldrich

Argininamide 5 mM – Manganese chloride 5 mM – Samarium(III) chloride 1 mM – Sodium cacodylate Buffer pH 6.0; 50 mM – 2-Methyl-2,4-pentanediol 30% solution

Synonym(s):

Crystallization solution 26/Kit-No 18839

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

form

solution

reaction suitability

reagent type: catalyst
core: samarium

pH

6.0±0.2 ( in neat)

storage temp.

2-8°C

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Carc. 1B - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laura C Kim et al.
Molecular cancer research : MCR, 18(11), 1675-1684 (2020-08-18)
Non-small cell lung cancer (NSCLC) is characterized by genomic alterations, yet a targetable mutation has not been discovered in nearly half of all patients. Recent studies have identified amplification of RICTOR, an mTORC2-specific cofactor, as a novel actionable target in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service