Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

91479

Sigma-Aldrich

6′-Sialyl-N-acetyllactosamine sodium salt

Na salt, ≥80% (TLC)

Synonym(s):

NeuAc alpha,6Gal beta1,4GIcNAc

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H42N2O19 · xNa+
CAS Number:
Molecular Weight:
674.60 (free acid basis)
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

assay

≥80% (TLC)

form

powder

Storage temp.

room temp

SMILES string

CC(=O)N[C@H]1C(O)O[C@H](CO)[C@@H](O[C@@H]2O[C@H](CO[C@]3(C[C@H](O)[C@@H](NC(C)=O)[C@@H](O3)[C@H](O)[C@H](O)CO)C(O)=O)[C@H](O)[C@H](O)[C@H]2O)[C@@H]1O

InChI

1S/C25H42N2O19/c1-7(30)26-13-9(32)3-25(24(40)41,46-21(13)15(34)10(33)4-28)42-6-12-16(35)18(37)19(38)23(44-12)45-20-11(5-29)43-22(39)14(17(20)36)27-8(2)31/h9-23,28-29,32-39H,3-6H2,1-2H3,(H,26,30)(H,27,31)(H,40,41)/t9-,10+,11+,12+,13+,14+,15+,16-,17+,18-,19+,20+,21+,22?,23-,25-/m0/s1

Inchi Key

RPSBVJXBTXEJJG-RAMSCCQBSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

6′-Sialyl-N-acetyllactosamine is an oligosaccharide with broad tissue distribution that is involved in multiple biological functions such as immunity and colon cancer progression. Certain strains of influenza bind with high affinity to 6′-Sialyl-N-acetyllactosamine.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service