Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

90911

Supelco

ZnAF-2 DA

~90% (HPCE)

Synonym(s):

6-{2-[Bis(2-pyridylmethyl)amino]ethylamino}fluorescein diacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H32N4O7
CAS Number:
Molecular Weight:
656.68
UNSPSC Code:
12352200
PubChem Substance ID:

assay

~90% (HPCE)

form

crystals

fluorescence

λex 492 nm; λem 515 nm in PBS, Zn2+, esterase

storage temp.

2-8°C

SMILES string

CC(=O)Oc1ccc2c(Cc3cc(OC(C)=O)ccc3C24OC(=O)c5ccc(NCCN(Cc6ccccn6)Cc7ccccn7)cc45)c1

InChI

1S/C38H32N4O7/c1-24(43)46-29-10-13-32-35(20-29)48-36-21-30(47-25(2)44)11-14-33(36)38(32)34-19-26(9-12-31(34)37(45)49-38)41-17-18-42(22-27-7-3-5-15-39-27)23-28-8-4-6-16-40-28/h3-16,19-21,41H,17-18,22-23H2,1-2H3

InChI key

QRHJESKPYRPLIW-UHFFFAOYSA-N

Application

Suitable for the fluorimetric detection of Zn2+ (after esterase hydrolysis).

hcodes

Hazard Classifications

Aquatic Chronic 4

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service