Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

90852

Sigma-Aldrich

Lectin from Phaseolus vulgaris-Atto 550 conjugate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

description

carbohydrate specificity Oligosaccharide

form

powder

fluorescence

λex 554 nm; λem 576 nm in PBS

Storage temp.

−20°C

General description

Lectins are ubiquitous proteins or glycoproteins with a high and specific affinity to sugar residues. They allow a sensitive detection of cellular carbohydrates, revealing subtle alteration in glycosylation between otherwise indistinguishable cells.
Atto-dyes have very bright fluorescent signals and high photostability, which enable a direct one-step tissue-binding protocol. Time-consuming multistage amplification procedures are not required for Atto-dye lectin conjugates

find more information here

Analysis Note

free of unconjugated dye

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Smart-aggregation imaging for single molecule localization with SPAD cameras.
Gyongy, I.; et al.
arXiv (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service