Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

90702

Sigma-Aldrich

Tectomer 3-tailed, CH3C(CH2-NH-Gly7)3 · 3CF3CO2H

≥85% (HPLC)

Synonym(s):

N,N′,N′″-Tris(glycylglycylglycylglycylglycylglycylglycine)-tris(2-aminomethyl)ethane tetra(trifluoroacetate) salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C47H78N24O21 · 3C2HF3O2
Molecular Weight:
1657.34
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥85% (HPLC)

form

powder

storage temp.

2-8°C

SMILES string

OC(=O)C(F)(F)F.OC(=O)C(F)(F)F.OC(=O)C(F)(F)F.OC(=O)C(F)(F)F.CC(CNC(=O)CNC(=O)CNC(=O)CNC(=O)CNC(=O)CNC(=O)CNC(=O)CN)(CNC(=O)CNC(=O)CNC(=O)CNC(=O)CNC(=O)CNC(=O)CNC(=O)CN)CNC(=O)CNC(=O)CNC(=O)CNC(=O)CNC(=O)CNC(=O)CNC(=O)CN

InChI

1S/C47H78N24O21.4C2HF3O2/c1-47(23-69-44(90)20-66-41(87)17-63-38(84)14-60-35(81)11-57-32(78)8-54-29(75)5-51-26(72)2-48,24-70-45(91)21-67-42(88)18-64-39(85)15-61-36(82)12-58-33(79)9-55-30(76)6-52-27(73)3-49)25-71-46(92)22-68-43(89)19-65-40(86)16-62-37(83)13-59-34(80)10-56-31(77)7-53-28(74)4-50;4*3-2(4,5)1(6)7/h2-25,48-50H2,1H3,(H,51,72)(H,52,73)(H,53,74)(H,54,75)(H,55,76)(H,56,77)(H,57,78)(H,58,79)(H,59,80)(H,60,81)(H,61,82)(H,62,83)(H,63,84)(H,64,85)(H,65,86)(H,66,87)(H,67,88)(H,68,89)(H,69,90)(H,70,91)(H,71,92);4*(H,6,7)

InChI key

VGQOKBWUNMAVGY-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service