Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

90592

Sigma-Aldrich

5-(Bromomethylene)-2(5H)-furanone

≥95.0% (HPLC)

Synonym(s):

Furanone C-56

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H3BrO2
CAS Number:
Molecular Weight:
174.98
Beilstein/REAXYS Number:
20240194
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

assay

≥95.0% (HPLC)

form

crystals

mp

82-86 °C

storage temp.

2-8°C

SMILES string

O=C1C=C/C(O1)=C\Br

InChI

1S/C5H3BrO2/c6-3-4-1-2-5(7)8-4/h1-3H/b4-3+

InChI key

LKFZLSZOWSMZDX-ONEGZZNKSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B X V Quecan et al.
Frontiers in microbiology, 10, 867-867 (2019-05-21)
Quorum sensing (QS) regulates bacterial gene expression and studies suggest quercetin, a flavonol found in onion, as a QS inhibitor. There are no studies showing the anti-QS activity of plants containing quercetin in its native glycosylated forms. This study aimed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service