Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

90308

Sigma-Aldrich

deGlycIT MicroSpin

deglycosylates up to 0.5 mg IgG, 5 columns

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204

form

suspension

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

5 x 0.1 ml settled resin of endoglycosidase IgGZERO (EndoS) immobilized on highly crosslinked agarose. Agarose resin is suspended in 20% ethanol/water (V/V)

Application

DeGlycIT MicroSpin columns (deglycosylates up to 0.5 mg IgG, 5 columns) can be used for antibody characterization.

Biochem/physiol Actions

DeGlycIT MicroSpin columns comprise of pre-filled endoglycosidase IgGZERO (EndoS) enzyme covalently coupled to highly cross-linked agarose. IgGZERO is a bacterial enzyme isolated from Streptococcus pyogenes, which possess hydrolytic activity specific for IgG bound glycans and cleaves bound IgG from IgG Fc receptors without damaging the native cells.[1]

Unit Definition

1 ml of settled enzyme immobilized agarose resin removes 95% of 5 mg of human IgG Fc glycans in 15 minutes at 25°C, pH 7.2 as monitored by SDS-PAGE.

Analysis Note

Enzymatic activity: corresponds to requirements

pictograms

Flame

signalword

Danger

hcodes

Hazard Classifications

Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

57.2 °F - closed cup

flash_point_c

14.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michele S Y Tan et al.
PloS one, 15(1), e0227341-e0227341 (2020-01-11)
Clan CA cysteine proteases, also known as papain-like proteases, play important roles throughout the malaria parasite life cycle and are therefore potential drug targets to treat this disease and prevent its transmission. In order to study the biological function of
Jonathan J Goodfellow et al.
Journal of the American Chemical Society, 134(19), 8030-8033 (2012-05-04)
Protein endoglycosidases are useful for biocatalytic alteration of glycans on protein surfaces, but the currently limited selectivity of endoglycosidases has prevented effective manipulation of certain N-linked glycans widely found in nature. Here we reveal that a bacterial endoglycosidase from Streptococcus

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service