NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, Muntjac skin, Fibroblast
tube of 5 μg 90112813-DNA-5UG pkg of vial of cells 90112813-1VL
growth mode
Adherent
karyotype
2n = 7
morphology
Fibroblast
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
Cell Line Origin
Muntjac skin
Cell Line Description
Derived from a skin biopsy of an adult male muntjac. The cells have a diploid chromosome number of 7 in the male and 6 in the female, amongst the lowest determined for a mammalian species. Recent tests indicate the cell line is positive for BVDV, both for antigen and infectious virus. Susceptible to vesicular stomatitis (Indiana strain), herpes simplex and vaccinia viruses.
Application
Chromosome studies
Packaging
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.
Split sub-confluent cultures (70-80%) 1:2 to 1:4 i.e. seeding at 2-4x10,000 cells/cm2using 0.25% trypsin/EDTA; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
Science (New York, N.Y.), 168(3937), 1364-1366 (1970-06-12)
The Indian muntjac (Muntiacus muntjak) has a diploid chromosome number of 7 in the male and 6 in the female, the lowest number yet described in a mammal. Its near relative, Reeve's muntjac (Muntiacus reevesi) has a diploid number of
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.