Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

89915

Sigma-Aldrich

2-[(N-Dansyl-biocytin)amido]ethyl-methanethiosulfonate

≥95% (coupling rate)

Synonym(s):

MTS-DB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H46N6O7S4
Molecular Weight:
742.99
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥95% (coupling rate)

form

solid

shelf life

limited shelf life, expiry date on the label

fluorescence

λem 560 nm±15 nm

storage temp.

−20°C

SMILES string

CN(C)c1cccc2c(cccc12)S(=O)(=O)NC(CCCCNC(=O)CCCC[C@@H]3SC[C@@H]4NC(=O)N[C@H]34)C(=O)NCCSS(C)(=O)=O

InChI

1S/C31H46N6O7S4/c1-37(2)25-13-8-11-22-21(25)10-9-15-27(22)48(43,44)36-23(30(39)33-18-19-46-47(3,41)42)12-6-7-17-32-28(38)16-5-4-14-26-29-24(20-45-26)34-31(40)35-29/h8-11,13,15,23-24,26,29,36H,4-7,12,14,16-20H2,1-3H3,(H,32,38)(H,33,39)(H2,34,35,40)/t23?,24-,26-,29-/m0/s1

InChI key

VGMGNIVELCIONM-UCIULOAQSA-N

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service