Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

89782

Sigma-Aldrich

Polyethylene glycol solution

BioUltra, 4,000, ~50% in H2O

Synonym(s):

PEG solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

Molecular Biology

product line

BioUltra

form

solution

concentration

~50% in H2O

impurities

DNases, none detected
RNases, none detected
insoluble matter, none detected
phosphatases, none detected
proteases, none detected

pH

5.0-6.5 (25 °C)

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤200 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤200 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

UV absorption

λ: 260 nm Amax: 0.6
λ: 280 nm Amax: 0.3

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

PEG is used as a fusogen to obtain hybridomas for monoclonal antibody production. Induces cell hybridization.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sondra T Bland et al.
Brain, behavior, and immunity, 23(4), 492-497 (2009-06-03)
Glial activation has recently been discovered to modulate several effects of morphine, including analgesia, tolerance, and dependence. The present studies extend this line of investigation by exploring whether glial activation may also affect extracellular levels of dopamine (DA) in the
Wanwisa Dejnirattisai et al.
The Journal of infectious diseases, 203(12), 1775-1783 (2011-05-25)
Dengue virus receptors are relatively poorly characterized, but there has been recent interest in 2 C-type lectin molecules, dendritic cell-specific intercellular adhesion molecule 3 (ICAM-3)-grabbing nonintegrin (DC-SIGN) and its close homologue liver/lymph node-specific ICAM-3-grabbing integrin (L-SIGN), which can both bind
Sergei A Filichkin et al.
Genome research, 20(1), 45-58 (2009-10-28)
Alternative splicing can enhance transcriptome plasticity and proteome diversity. In plants, alternative splicing can be manifested at different developmental stages, and is frequently associated with specific tissue types or environmental conditions such as abiotic stress. We mapped the Arabidopsis transcriptome
Seyeon Bae et al.
Immune network, 11(3), 175-181 (2011-08-24)
CM1 (centrocyte/-blast marker 1) was defined by a mAb against concanavalin A (Con A) activated PBMC. It is expressed in germinal center of human tonsil and on the surface of activated PBMC as well as cancer cells. Recently, increased productions
Mark R Hutchinson et al.
Brain, behavior, and immunity, 23(2), 240-250 (2008-10-22)
Morphine-induced glial proinflammatory responses have been documented to contribute to tolerance to opioid analgesia. Here, we examined whether drugs previously shown to suppress glial proinflammatory responses can alter other clinically relevant opioid effects; namely, withdrawal or acute analgesia. AV411 (ibudilast)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service