Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

89137

Sigma-Aldrich

Lipase, immobilized in Sol-Gel-AK on pumice from Candida antarctica

≥1.5 U/g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204

form

beads

specific activity

≥1.5 U/g

Storage temp.

2-8°C

Application

Lipases are used industrially for the resolution of chiral compounds and the transesterification production of biodiesel.

Biochem/physiol Actions

Lipases catalyze the hydrolysis of triacylglycerols into glycerol and free fatty acids.

Unit Definition

1 U corresponds to the amount of enzyme which liberates 1 μmol butyric acid per minute at pH 8.0 and 40°C (tributyrin, Cat. No. 91010, as substrate)

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service