Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

88438

Sigma-Aldrich

TRIS-HCl pH 8.5; 0.1M – Ethanol 20% (v/v) solution

Synonym(s):

Crystallization solution 44/Kit-No 70437

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

form

solution

pH

8.5±0.2 ( in neat)

density

0.98 g/mL at 20 °C

storage temp.

2-8°C

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

105.8 °F - closed cup

flash_point_c

41 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sunderajhan Sekar et al.
Nutrition (Burbank, Los Angeles County, Calif.), 54, 144-152 (2018-06-01)
Obesity is a known strong risk factor for the onset of knee osteoarthritis and is often accompanied by dysregulated lipid metabolism with elevated levels of free fatty acids such as saturated fatty acids (SFAs). The purpose of this study was

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service