Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

86696

Sigma-Aldrich

Tetraethylene glycol monooctadecyl ether

BioXtra, ≥99.0% (GC)

Synonym(s):

C18E4, Octadecyl tetraethylene glycol ether, Octadecyltetraglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)17(OCH2CH2)4OH
CAS Number:
Molecular Weight:
446.70
Beilstein/REAXYS Number:
1884041
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:

description

non-ionic

product line

BioXtra

assay

≥99.0% (GC)

form

solid

mol wt

446.70 g/mol

mp

44-46 °C

cation traces

Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cr: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤50 mg/kg
Mg: ≤50 mg/kg
Mn: ≤50 mg/kg
Na: ≤50 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

CCCCCCCCCCCCCCCCCCOCCOCCOCCOCCO

InChI

1S/C26H54O5/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-19-28-21-23-30-25-26-31-24-22-29-20-18-27/h27H,2-26H2,1H3

InChI key

HEZHDXIJQPSERX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service