Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

85861

Sigma-Aldrich

Water : ethanol : acetic acid 6:3:1

Synonym(s):

Acetic acid : ethanol : water 1:3:6

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.26

form

liquid

dilution

(for analytical testing)

refractive index

n20/D 1.360

Application

Solution for fixation of proteins in electrophoresis gels after staining with Ruthenium complexes (e.g. Tris(bathophenanthrolinedisulfonate)ruthenium (II) sodium salt (03038))

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

55.4 °F

flash_point_c

13 °C

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service