Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

85570C

LONG® EGF human

recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

Synonym(s):

Epidermal Growth Factor, EGF

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352200

recombinant

expressed in E. coli

description

for research or for futher manufacturing use

form

lyophilized powder

availability

not available in Japan

technique(s)

cell culture | mammalian: suitable

storage temp.

2-8°C

Application

LONG®EGF is a recombinant analog of human epidermal growth factor (EGF) that provides a potent and cost-effective mitogen for cell culture applications. LONG®EGF is ideal for both research and large-scale culture systems utilizing serum-free or low-level serum applications. It is effective in a variety of fibroblast and endothelial cell types and also acts synergistically with LONG®R3IGF-I (Catalog No. 85580C) to produce extremely potent results in cell culture. LONG®EGF is produced in a patented E. coli expression system free of animal-derived components.

Analysis Note

The biological activity is measured by the stimulation of protein synthesis in L6 myoblasts

Other Notes

This product should be purchased through SAFC Biosciences by contacting their customer service at the following phone numbers: in North America, 1-800-255-6032 or +1 913-469-5580; in Europe, +44 (0) 1264-333311; and in Asia-Pacific (including Australia and Japan), +61 (0)3-9362-4500 or 1-800-200-404.

Legal Information

Available in USA, Canada, and Europe. Please check with your local supplier regarding availability and restrictions in other countries.
LONG is a registered trademark of Repligen Sweden AB

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoni Li et al.
The Journal of biological chemistry, 288(25), 18121-18133 (2013-05-08)
Emerging evidence indicates that the miR-23a/24-2/27a cluster may possess a causal role in mammary tumorigenesis and function as a novel class of oncogenes. However, the regulatory mechanism of the miR-23a/24-2/27a cluster in mammary carcinoma cell invasion and migration is still
C-J Kao et al.
Oncogene, 33(19), 2495-2503 (2013-06-04)
Src tyrosine kinase (Src) is implicated in the development of bone metastasis and castration resistance of prostate cancer. Src inhibitors are currently being tested in clinical trials for such diseases. Understanding the molecular and cellular actions of Src inhibitors holds
Chi-Yuan Chen et al.
Cancer research, 73(13), 4009-4019 (2013-05-24)
Tid1 (DNAJA3), a DnaJ cochaperone, may promote degradation of oncogenic kinases. Tid1 has 2 isoforms, Tid1-L and Tid1-S, that may function differently. In this study, we investigated the role of the Tid1 isoforms in regulating EGF receptor (EGFR) signaling and
Yong Zheng et al.
Nature, 499(7457), 166-171 (2013-07-13)
Cell-surface receptors frequently use scaffold proteins to recruit cytoplasmic targets, but the rationale for this is uncertain. Activated receptor tyrosine kinases, for example, engage scaffolds such as Shc1 that contain phosphotyrosine (pTyr)-binding (PTB) domains. Using quantitative mass spectrometry, here we
Olivia I Kober et al.
American journal of physiology. Gastrointestinal and liver physiology, 306(7), G582-G593 (2014-02-08)
Intestinal homeostasis is maintained by a hierarchy of immune defenses acting in concert to minimize contact between luminal microorganisms and the intestinal epithelial cell surface. The intestinal mucus layer, covering the gastrointestinal tract epithelial cells, contributes to mucosal homeostasis by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service