Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

85447

Sigma-Aldrich

Sissotrin

~95% (TLC)

Synonym(s):

Biochanin A 7-O-β-D-glucopyranoside, Sissostrin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H22O10
CAS Number:
Molecular Weight:
446.40
Beilstein/REAXYS Number:
1302648
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

assay

~95% (TLC)

form

powder

SMILES string

COc1ccc(cc1)C2=COc3cc(O[C@@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O)cc(O)c3C2=O

InChI

1S/C22H22O10/c1-29-11-4-2-10(3-5-11)13-9-30-15-7-12(6-14(24)17(15)18(13)25)31-22-21(28)20(27)19(26)16(8-23)32-22/h2-7,9,16,19-24,26-28H,8H2,1H3/t16-,19-,20+,21-,22-/m1/s1

Inchi Key

LFEUICHQZGNOHD-RECXWPGBSA-N

General description

Flavonoid glycoside from Sophora japonica (Japanese pagoda tree), Cicer arietinum (chickpea), and other plant sources.

Biochem/physiol Actions

Sissotrin in the leaf exudate of chickpea inactivates nucleopolyhedroviruses (such as baculovirus) which can be used as biological pesticides..

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Max Norman Tandrup Lambert et al.
PloS one, 12(6), e0176590-e0176590 (2017-06-08)
Natural estrogen decline leads to vasomotor symptoms (VMS). Hormone therapy alleviates symptoms but increases cancer risk. Effective treatments against VMS with minimal cancer risks are needed. We investigate the effects of a highly bioavailable aglycone rich Red Clover isoflavone treatment

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service