Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

84434

Sigma-Aldrich

Hydrochloric acid solution

BioXtra, ~0.1 M in H2O

Synonym(s):

Hydrogen chloride solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HCl
CAS Number:
Molecular Weight:
36.46
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

BioPerformance Certified

vapor pressure

190 mmHg ( 25 °C)

product line

BioXtra

form

liquid

concentration

0.096-0.104 M (alkalimetric)
~0.1 M in H2O

impurities

insoluble matter, passes filter test

pH

1 (25 °C, 0.1 M in H2O)

bp

100 °C (lit.)

density

1.00 g/mL at 20 °C

anion traces

sulfate (SO42-): ≤10 mg/kg

cation traces

Al: ≤1 mg/kg
As: ≤0.1 mg/kg
Ba: ≤1 mg/kg
Bi: ≤1 mg/kg
Ca: ≤5 mg/kg
Co: ≤1 mg/kg
Cr: ≤1 mg/kg
Cu: ≤1 mg/kg
Fe: ≤1 mg/kg
K: ≤20 mg/kg
Li: ≤1 mg/kg
Mg: ≤1 mg/kg
Mn: ≤1 mg/kg
Mo: ≤1 mg/kg
Na: ≤20 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/kg
Sr: ≤1 mg/kg
Zn: ≤1 mg/kg

λ

neat

UV absorption

λ: 260 nm Amax: 0.01
λ: 280 nm Amax: 0.01

storage temp.

2-8°C

SMILES string

Cl

InChI

1S/ClH/h1H

InChI key

VEXZGXHMUGYJMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Douglas M McMillan et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 40(7), 1804-1812 (2015-01-30)
CYP2D metabolically activates codeine to morphine, which is required for codeine analgesia. Permeability across the blood-brain barrier, and active efflux, suggests that initial morphine in the brain after codeine is due to brain CYP2D metabolism. Human CYP2D is higher in
Marta Kolodziejczak et al.
ACS chemical neuroscience, 6(7), 1219-1230 (2015-04-11)
Maturation of functional neuronal circuits during central nervous system development relies on sophisticated mechanisms. First, axonal and dendritic growth should reach appropriate targets for correct synapse elaboration. Second, pruning and neuronal death are required to eliminate redundant or inappropriate neuronal
Davide R Ceratti et al.
Advanced materials (Deerfield Beach, Fla.), 27(34), 4958-4962 (2015-07-23)
A new, simple bi-phasic dip-coating method is developed. This method is considered as a great improvement of the technique for research, development and production, since expensive, rare, harmful, or time-evolving solutions can now be easily deposited on large surfaces and
Xuan Zhang et al.
Nanomedicine : nanotechnology, biology, and medicine, 11(6), 1365-1375 (2015-04-19)
Intratumoral injection of ultra-small gold nanoparticles (AuNPs) conjugated to doxorubicin (Au-Dox) is effective against both murine B16 and human SK-MEL-28 tumors in mice. Au-Dox suppresses growth of B16 tumors in immunocompetent mice by >70% for at least 19 days. In
Rebecca J Ford et al.
The Biochemical journal, 468(1), 125-132 (2015-03-06)
Metformin is the mainstay therapy for type 2 diabetes (T2D) and many patients also take salicylate-based drugs [i.e., aspirin (ASA)] for cardioprotection. Metformin and salicylate both increase AMP-activated protein kinase (AMPK) activity but by distinct mechanisms, with metformin altering cellular

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service