83853
Ribonucleic acid, transfer from baker′s yeast (S. cerevisiae)
~20 A260/mg
Synonym(s):
Transfer RNA, tRNA
About This Item
Recommended Products
grade
Molecular Biology
form
solid
concentration
~20 A260/mg
solubility
H2O: 10 mg/mL, clear, light yellow
shipped in
wet ice
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
General description
Application
related product
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service