Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

83756

Sigma-Aldrich

Fluorescent red 610

suitable for fluorescence

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H42N3NaO9S
CAS Number:
Molecular Weight:
667.75
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32

form

powder

solubility

DMSO: soluble

fluorescence

λex 610 nm; λem 645 nm in 0.1 M phosphate pH 7.0

suitability

suitable for fluorescence

Storage temp.

2-8°C

InChI

1S/C32H43N3O9S.Na/c1-6-33(7-2)22-13-15-24-26(32(3,4)5)21-23(44-27(24)20-22)14-16-25-29(38)34(17-10-8-9-12-28(36)37)31(40)35(30(25)39)18-11-19-45(41,42)43;/h13-16,20-21H,6-12,17-19H2,1-5H3,(H,36,37)(H,41,42,43);/q;+1/p-1

Inchi Key

HMAWMSKJKAVASI-UHFFFAOYSA-M

General description

Fluorescent red 610 is a fluorescent dye especially well suited for excitation by He-Ne-lasers (at 594 nm). Fluorescent red 610 shows strong fluorescence (molar absorption 81 000 M-1cm-1)

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service