Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

83390

Sigma-Aldrich

Quercetin-3-D-xyloside

≥97.0% (HPLC)

Synonym(s):

Reinutrin, Reynoutrin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H18O11
CAS Number:
Molecular Weight:
434.35
Beilstein/REAXYS Number:
6023007
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.47

assay

≥97.0% (HPLC)

form

crystals

SMILES string

O[C@@H]1COC(OC2=C(Oc3cc(O)cc(O)c3C2=O)c4ccc(O)c(O)c4)[C@H](O)[C@H]1O

InChI

1S/C20H18O11/c21-8-4-11(24)14-13(5-8)30-18(7-1-2-9(22)10(23)3-7)19(16(14)27)31-20-17(28)15(26)12(25)6-29-20/h1-5,12,15,17,20-26,28H,6H2/t12-,15+,17-,20?/m1/s1

Inchi Key

PZZRDJXEMZMZFD-DUYPVGENSA-N

Other Notes

Flavonol glycoside present in quince, pear, and apple[1][2] and cranberry. Shows potent free-radical scavenging and antioxidant activity in vivo.[3]

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W. Oleszek et al.
Journal of Agricultural and Food Chemistry, 36, 430-430 (1988)
Xiaojun Yan et al.
Journal of agricultural and food chemistry, 50(21), 5844-5849 (2002-10-03)
Polyphenolic compounds in cranberries have been investigated to determine their role in protection against cardiovascular disease and some cancers. Extracts of whole fruit were assayed for radical-scavenging activity and tumor growth inhibition using seven tumor cell lines. Selective inhibition of
S. Burda et al.
Journal of Agricultural and Food Chemistry, 38, 945-945 (1990)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service