Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

80925

Sigma-Aldrich

Ammonium phosphate monobasic 0.2M – TRIS-HCl pH 8.5; 0.1M – 2-Methyl-2,4-pentanediol 50% (v/v) solution

Synonym(s):

Crystallization solution 43/Kit-No 70437

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

form

solution

pH

8.5±0.2 ( in neat)

density

1.01 g/mL at 20 °C

storage temp.

2-8°C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service