Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

80859

Sigma-Aldrich

Potassium tert-butoxide

Synonym(s):

Potassium tert-butylate, Potassium t-butoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COK
CAS Number:
Molecular Weight:
112.21
Beilstein/REAXYS Number:
3556712
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

vapor pressure

1 mmHg ( 220 °C)

form

solid

mp

256-258 °C (dec.) (lit.)

SMILES string

[K+].CC(C)(C)[O-]

InChI

1S/C4H9O.K/c1-4(2,3)5;/h1-3H3;/q-1;+1

InChI key

LPNYRYFBWFDTMA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yong Guo et al.
Organic & biomolecular chemistry, 7(8), 1716-1722 (2009-04-04)
Alpha-fluoro-alpha-aryl-ketones were synthesized by the Pd-catalyzed cross-coupling of aryl bromides with either alpha-fluoroketones or their corresponding silyl enol ethers. The direct arylation with an alpha-fluoroketone requires a strong base, such as potassium tert-butoxide, and under these conditions the presence of
Brian C Anderson et al.
Biomaterials, 23(22), 4345-4352 (2002-09-11)
Several homopolymers and copolymers of 2-(diethylamino)ethyl methacrylate (DEAEM) and poly(ethylene glycol) methyl ether methacrylate (PEGMEM) were synthesized using anionic polymerization initiated by potassium t-butoxide. The polymers were characterized by average molecular weight, polydispersity and monomeric unit composition. A very narrow
Astrid Beyer et al.
Organic letters, 14(15), 3948-3951 (2012-07-17)
Potassium tert-butoxide-mediated intramolecular α-arylations of fluoro- and chloro-substituted anilides provide oxindoles in DMF at 80 °C. In this manner, diversely substituted products have been obtained in moderate to high yields.
Luís S Monteiro et al.
Amino acids, 43(4), 1643-1652 (2012-02-22)
Recently we reported the use of a sequence of alkylation and dehydration methodologies to obtain N-ethyl-α, β-dehydroamino acid derivatives. The application of this N-alkylation procedure to several methyl esters of β,β-dibromo and β-bromo, β-substituted dehydroamino acids protected with standard amine
Wei Liu et al.
Chemical communications (Cambridge, England), 49(29), 2983-2985 (2013-03-07)
Simple and cheap alcohols can promote the direct arylation of unactivated arenes with aryl iodides and bromides in the presence of potassium tert-butoxide. This transition-metal-free aromatic C-H transformation offers a cheap and easy practical way to synthesize biaryls under mild

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service