Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

80837

Sigma-Aldrich

D-Fuconic acid lithium salt

≥98.5% (TLC)

Synonym(s):

6-Deoxy-D-galactonic acid lithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6 · xLi+
CAS Number:
Molecular Weight:
180.16 (free acid basis)
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

Quality Level

assay

≥98.5% (TLC)

form

powder

storage temp.

2-8°C

SMILES string

C[C@@H](O)[C@H](O)[C@H](O)[C@@H](O)C(O)=O

InChI

1S/C6H12O6/c1-2(7)3(8)4(9)5(10)6(11)12/h2-5,7-10H,1H3,(H,11,12)/t2-,3+,4+,5-/m1/s1

InChI key

NBFWIISVIFCMDK-MGCNEYSASA-N

Biochem/physiol Actions

Metabolite of D-fucose metabolism, enzyme substrate for D-fuconate dehydratase.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D-Galactonate/D-fuconate dehydratase.
R L Anderson et al.
Methods in enzymology, 90 Pt E, 299-302 (1982-01-01)
D-Fucose metabolism in a pseudomonad. II. Oxidation of D-fucose to D-fucono- -lactone by an L-arabino-aldose dehydrogenase and hydrolysis of the lactone by a lactonase.
A S Dahms et al.
The Journal of biological chemistry, 247(7), 2228-2232 (1972-04-10)
D-Fucose metabolism in a pseudomonad. 3. Conversion of D-fuconate to 2-keto-3-deoxy-D-fuconate by a dehydratase.
A S Dahms et al.
The Journal of biological chemistry, 247(7), 2233-2237 (1972-04-10)
Wen Shan Yew et al.
Biochemistry, 45(49), 14582-14597 (2006-12-06)
Many members of the mechanistically diverse enolase superfamily have unknown functions. In this report we use both genome (operon) context and screening of a library of acid sugars to assign the L-fuconate dehydratase (FucD) function to a member of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service