Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

80720

Sigma-Aldrich

cis,cis-Farnesol

≥95.0% (GC)

Synonym(s):

(2Z,6Z)-3,7,11-Trimethyl-2,6,10-dodecatrien-1-ol, 2-cis,6-cis-Farnesol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H26O
CAS Number:
Molecular Weight:
222.37
Beilstein/REAXYS Number:
1723036
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

assay

≥95.0% (GC)

form

liquid

storage temp.

−20°C

SMILES string

C\C(C)=C\CC\C(C)=C/CC\C(C)=C/CO

InChI

1S/C15H26O/c1-13(2)7-5-8-14(3)9-6-10-15(4)11-12-16/h7,9,11,16H,5-6,8,10,12H2,1-4H3/b14-9-,15-11-

InChI key

CRDAMVZIKSXKFV-FBXUGWQNSA-N

Biochem/physiol Actions

Farnesol is an acyclic sesquiterpene alcohol, with four possible isomers, the E/E-isomer being the most common in nature. It was found to have different biological functions, for example to act as a quorum-sensing molecule to suppress filamentation in the fungus Candida albicans; and to have antibacterial effects against Staphylococcus aureus, including promotion of potassium ion leakage. Its derivatives are involved in the a-factor mating peptide of the dodecapeptide pheromone found in Saccharomyces cerevisiae. The biological activity of farnesol and FPP analogues are investigated in cancer research.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Xie et al.
The Journal of organic chemistry, 65(25), 8552-8563 (2000-12-12)
The a-factor of Saccharomyces cerevisiae is a dodecapeptide pheromone (YIIKGVFWDPAC(Farnesyl)-OCH(3), 1), in which post-translational modification with a farnesyl isoprenoid and carboxymethyl group is required for full biological activity. This peptide has been used as a model system to explore the
R J Hohl et al.
Lipids, 33(1), 39-46 (1998-02-21)
This investigation compares the effects of three farnesyl pyrophosphate analogs on selected aspects of isoprenoid metabolism. E,E-alpha-Hydroxyfarnesylphosphonate was prepared by an improved variation on a literature synthesis, which also gave access to the new Z,E-alpha-hydroxyfarnesyl- and alpha-hydroxygeranylphosphonates. A striking find
J Downward
Current opinion in genetics & development, 8(1), 49-54 (1998-04-08)
Activated Ras proteins have either positive or negative effects on the regulation of apoptosis depending on cell type and other factors. In part, this is due to the ability of Ras to control directly multiple effector pathways, including PI3-kinase, which
José S Yu et al.
Organic letters, 7(22), 4803-4806 (2005-10-21)
[structure: see text] The four olefin stereoisomers of farnesol have been synthesized from readily available nerylacetone or commercial geranylacetone. A new variation on the use of beta-oxido ylides favored the (2Z)-stereoisomers, whereas the (2E)-isomers were obtained through a classical Horner-Wadsworth-Emmons
Yoshihiro Inoue et al.
FEMS microbiology letters, 237(2), 325-331 (2004-08-24)
The study was made of the antibacterial effects of three terpene alcohols on Staphylococcus aureus, focusing on the leakage of K+ ions and toxicity over time. The leakage of K+ ions was monitored continuously with a K+-electrode. Our results suggested

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service