Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

80701

Sigma-Aldrich

Crystallization Kit for DNA

Synonym(s):

DNA Crystallization Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12171500

storage temp.

2-8°C

General description

The Crystallization Kit for DNA is an empirical screening method for the direct determination of suitable crystallization conditions for nucleic acids.

The kit is developed according to the described method of Nowakowski et al.[1], Baeyens, Jancarik and Holbrook[2] These ready-made solutions represent a screen of conditions, from which many were observed to produce crystals from DNA oligomers and transfer RNA. This screen provides high efficiency in crystallization of RNA oligomers. Thus it provides a completion of our crystallization kit for DNA.

The Crystallization Kit for DNA consists of 48 different formulations covering a range of different buffers (pH 6.0-8.0), different types of precipitants (esp. PEG 400 and MPD), and inorganic salts including magnesium salts. All solutions are sterile filtered using 0.22 micron filters. The kit contains 10 ml of each formulation, but all solutions are available separately as 100 ml bottles. Larger quantities are available on request.

Features and Benefits

  • Comprehensive, efficient screening
  • Extremely pure, sterile reagents
  • Prefabricated reagents and observation sheets
  • Low incidence of undesirable solubility effects

Other Notes

Every component can be ordered separately in pack size of 100 mL, but inquiries for different sizes are also welcome.
The Kit contains 48 components with 10 mL each.

Kit Components Only

Product No.
Description

  • Cacodylate pH 6.5; 0.05 M − Magnesium chloride 5 mM − Cobalt(III)-hexamine 2.5 mM solution

  • Cacodylate pH 6.5; 0.05 M − Magnesium chloride 10 mM − Sodium citrate 200 mM − Isopropanol 5% solution

  • Cacodylate pH 6.5; 0.05 M − Magnesium chloride 15 mM − Spermine 10.0 mM − Lithium sulfate 2.0 M solution

  • Cacodylate pH 6.5; 0.05 M − Magnesium chloride 10 mM − Spermine 1.5 mM − Ammonium sulfate 3.0 solution

  • Cacodylate pH 6.0; 0.05 M − Calcium acetate 200 mM − Sodium chloride 2.5 M solution

  • Cacodylate pH 6.0; 0.05 M − Calcium acetate 200 mM − Cobalt(III)-hexamine 1.0 mM − Lithium chloride 2.0 M solution

  • TRIS pH 7.5; 0.05 M − Magnesium chloride 200 mM − Sodium chloride 2.5 M solution

  • Cacodylate pH 6.0; 0.05 M − Magnesium chloride 200 mM − Potassium chloride 2.5 M solution

  • Cacodylate pH 6.5; 0.05 M − Spermine 1.0 mM − Cobalt(III)-hexamine 2.0 mM − Calcium chloride 30 mM − Lithium chloride 2.0 M solution

See All (9)

Kit Components Also Available Separately

Product No.
Description
SDS

  • HEPES pH 7.5; 0.05 M − Magnesium chloride 80 mM − Spermine 2.5 mM solution

  • Cacodylate pH 6.0; 0.05 M − Magnesium chloride 18 mM − Spermine 2.25 mM − Copper sulfate 1 mM − Isopropanol 9% solution

  • Cacodylate pH 6.5; 0.05 M − Magnesium chloride 18 mM − Spermine 0.9 mM − Cobalt(III)-hexamine 1.8 mM − Isopropanol 9% solution

  • Cacodylate pH 6.5; 0.05 M − Magnesium chloride 18 mM − Spermine 2.25 mM − Isopropanol 9% solution

  • Cacodylate pH 7.0; 0.05 M − Magnesium chloride 18 mM − Spermine 2.25 mM − Cobalt(III)-hexamine 0.9 mM − 2-Methyl-2,4-pentanediol 4.5% solution

  • Cacodylate pH 6.5; 0.05 M − Magnesium chloride 36mM − Spermine 2.25 mM − PEG 400 5% solution

  • Sodium succinate pH 5.5; 0.05 M − Magnesium chloride 10 mM − Cobalt(III)-hexamine 2.0 mM − Isopropanol 10% solution

  • Cacodylate pH 6.0; 0.05 M − Magnesium chloride 20 mM − Spermine 1.0 mM − Ethanol 15% solution

  • Cacodylate pH 7.0; 0.05 M − Magnesium chloride 20 mM − Spermine 1.0 mM − Cobalt(III)-hexamine 1.0 mM − Ethanol 15% solution

  • Cacodylate pH 7.0; 0.05 M − Magnesium chloride 5 mM − Spermidine 1.0 mM − tert-Butanol 10% solution

  • Cacodylate pH 7.0; 0.05 M − Magnesium chloride 30 mM − Spermine 2.5 mM − PEG 400 5% solution

  • Cacodylate pH 6.5; 0.05 M − Magnesium chloride 100 mM − Cobalt(III)-hexamine 2.0 mM − Isopropanol 5% solution

  • TRIS pH 8.0; 0.05 M − Magnesium chloride 10 mM − Cobalt(III)-hexamine 1.0 mM − Ethanol 20% solution

  • HEPES pH 7.5; 0.05 M − Magnesium chloride 20 mM − Spermine 1.0 mM − PEG 8000 5% solution

  • Cacodylate pH 6.0; 0.05 M − Magnesium chloride 20 mM − Spermine 2.5 mM − PEG 4000 5% solution

  • Cacodylate pH 6.0; 0.05 M − Magnesium chloride 10 mM − Spermine 2.5 mM − Calcium chloride 5 mM − Isopropanol 10% solution

  • Cacodylate pH 7.0; 0.05 M − Magnesium chloride 9 mM − Spermine 2.25 mM − Cobalt(III)-hexamine 1.8 mM − Spermidine 0.9 mM − PEG 400 5% solution

  • Cacodylate pH 6.5; 0.05 M − Magnesium chloride 10 mM − Spermine 2.5 mM − Copper sulfate 1 mM − Isopropanol 10% solution

  • Cacodylate pH 6.0; 0.05 M − Magnesium chloride 20 mM − Spermine 1.0 mM − Calcium chloride 2 mM − 1,5 Hexanediol 10% solution

  • HEPES pH 7.5; 0.05 M − Magnesium chloride 15 mM − Spermidine 1.0 mM − Dioxan 10% solution

  • Cacodylate pH 6.0; 0.05 M − Magnesium chloride 15 mM − Spermine 3.0 mM − PEG 400 10% solution

  • Cacodylate pH 6.5; 0.05 M − Spermine 2.5 mM − Calcium chloride 18 mM − 2-Propanol 9% solution

  • Cacodylate pH 6.5; 0.05 M − Spermine 2.0 mM − Cobalt(III)-hexamine 1.0 mM − Calcium chloride 80 mM solution

  • Cacodylate pH 6.5; 0.05 M − Magnesium chloride 30 mM − Spermine 1.0 mM − Lithium sulfate 1.3 M solution

  • Cacodylate pH 6.0; 0.05 M − Calcium acetate 200 mM − Isopropanol 5% solution

  • Cacodylate pH 6.5; 0.05 M − Magnesium chloride 100 mM − Cobalt(III)-hexamine 1.0 mM − Ethanol 10% solution

  • Cacodylate pH 6.0; 0.05 M − Magnesium chloride 10 mM − Spermidine 2.5 mM − Sodium chloride 2.5 M solution

  • Cacodylate pH 6.5; 0.05 M − Magnesium chloride 20 mM − Spermine 1.0 mM − Ammonium sulfate 2.0 M solution

  • Cacodylate pH 6.5; 0.05 M − Magnesium chloride 15 mM − Spermidine 5.0 mM − Cobalt(III)-hexamine 1.0 mM − Sodium chloride 2.0 M solution

  • Cacodylate pH 6.5; 0.05 M − Magnesium chloride 200 mM − Sodium chloride 100 mM − PEG 1000 20% solution

  • TRIS pH 7.5; 0.05 M − Magnesium chloride 50 mM − Sodium tartrate 1.0 M solution

  • TRIS pH 8.0; 0.05 M – Magnesium chloride 200 mM – Ethanol 15% solution

  • Cacodylate pH 6.0; 0.05 M − Magnesium chloride 15 mM − Spermidine 5.0 mM − Lithium sulfate 2.0 M solution

  • Cacodylate pH 6.0; 0.05 M − Magnesium acetate 20 mM − Spermine 0.5 mM − Sodium chloride 100 mM − 2-Methyl-2,4-pentanediol 25% solution

  • Sodium succinate 0.05 M − Magnesium chloride 20 mM − Spermine 0.5 mM − Ammonium sulfate 3.0 solution

  • Cacodylate pH 6.5; 0.05 M − Cobalt(III)-hexamine 5 mM − Potassium chloride 2.5 M solution

  • Cacodylate pH 6.5; 0.05 M − Magnesium chloride 50 mM − Cobalt(III)-hexamine 2.0 mM − Lithium sulfate 1.5 M solution

  • Cacodylate pH 6.5; 0.05 M − Magnesium chloride 10 mM − Spermine 50 mM solution

See All (38)

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Nowakowski et al.
Acta crystallographica. Section D, Biological crystallography, 55(Pt 11), 1885-1892 (1999-10-26)
One of the most difficult steps in the X-ray crystallography of nucleic acids is obtaining crystals that diffract to high resolution. The choice of the nucleotide sequence has proven to be more important in producing high-quality crystals than the composition
K J Baeyens et al.
Acta crystallographica. Section D, Biological crystallography, 50(Pt 5), 764-767 (1994-09-01)
We have crystallized a variety of RNA oligonucleotides in a form suitable for X-ray diffraction studies using polyethylene glycol with a low-molecular-weight distribution (PEG 400) as the precipitant. Crystallization experiments on a set of 26 RNA oligomers ranging from eight

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service