Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

80472

Sigma-Aldrich

Atto 430LS-Streptavidin

suitable for fluorescence

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

form

powder

Quality Level

manufacturer/tradename

ATTO-TEC GmbH

fluorescence

λex 433 nm; λem 530 nm±10 nm in PBS, pH 7.4

suitability

suitable for fluorescence

Storage temp.

−20°C

Analysis Note

unconjugated dye ≤5% of total fluorescence

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service