80351
N-Methyl-cis-4-hydroxy-L-proline
≥98.0% (TLC)
Synonym(s):
(4S)-4-Hydroxy-1-methyl-L-proline, N-Me-cis-Hyp-OH, N-Methyl-L-allohydroxyproline
About This Item
Recommended Products
Quality Level
assay
≥98.0% (TLC)
form
powder
optical activity
[α]/D -68.5±2.5°, c = 1 in H2O
SMILES string
CN1C[C@@H](O)C[C@H]1C(O)=O
InChI
1S/C6H11NO3/c1-7-3-4(8)2-5(7)6(9)10/h4-5,8H,2-3H2,1H3,(H,9,10)/t4-,5-/m0/s1
InChI key
FMIPNAUMSPFTHK-WHFBIAKZSA-N
Biochem/physiol Actions
Packaging
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Sens. 1
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service