Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

80347

Sigma-Aldrich

Nano beads based on polyacrylonitrile, Chromeon 545-marked, coated with streptavidin

Synonym(s):

PAN-Chromeon 545, streptavidin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

form

beads

concentration

0.5%

average diameter

80-100 nm

fluorescence

λex 545 nm; λem 579 nm in H2O

storage temp.

2-8°C

Application

Fluorescent PAN (polyacrylnitrile) particles are specially suitable for energy transfer with external fluroescent dyes and embedded dyes.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service