Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

80091

Sigma-Aldrich

Lithium sulfate 0.17 M – TRIS hydrochloride pH 8.5; 0.085M – Polyethylen glycol 4000 25.5% (w/v) – Glycerol 15% (v/v) solution

Synonym(s):

Crystallization solution 17/Kit No 75403

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

form

liquid

pH

8.5±0.2 ( in neat)

density

1.10 g/mL at 20 °C

storage temp.

2-8°C

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eric Gyimah et al.
Chemosphere, 262, 128045-128045 (2020-11-14)
The vulnerability to environmental insults is heightened at early stages of development. However, the neurotoxic potential of bisphenol A (BPA) and bisphenol S (BPS) at developmental windows remains unclear. To investigate the mechanisms mediating the developmental neurotoxicity, zebrafish embryos were

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service