Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

80053

Sigma-Aldrich

Ammonium sulfate 0.16 M – Sodium acetate pH 4.6; 0.08M – Polyethylene glycol 4000 20% (w/v) – Glycerol 20% (v/v) solution

Synonym(s):

Crystallization solution 20/Kit No 75403

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

form

liquid

pH

4.6±0.2 ( in neat)

density

1.10 g/mL at 20 °C

storage temp.

2-8°C

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service