79974
Magnesium acetate 0.2M – Sodium cacodylate pH 6.5; 0.1M – 2-Methyl-2,4-pentanediol 30% (v/v) solution
Synonym(s):
Crystallization solution 21/Kit No 75403
About This Item
form
liquid
pH
6.5±0.2 ( in neat)
density
1.02 g/mL at 20 °C
storage temp.
2-8°C
signalword
Danger
Hazard Classifications
Aquatic Chronic 2 - Carc. 1B - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service