79971
N-(2-Acetamido)iminodiacetic acid pH 6.5; 0.1M – 2-Methyl-2,4-pentanediol 12% (v/v) solution
Synonym(s):
Crystallization solution 26/Kit-No 73513
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352200
form
solution
pH
6.5±0.2 ( in neat)
density
1.01 g/mL at 20 °C
storage temp.
2-8°C
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service