Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

79272

Sigma-Aldrich

D-Threonic acid lithium salt

≥98.5% (TLC)

Synonym(s):

(2S,3R)-2,3,4-Trihydroxybutyric acid lithium salt, Lithium-D-threonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O5 · xLi+
CAS Number:
Molecular Weight:
136.10 (free acid basis)
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

assay

≥98.5% (TLC)

form

powder

storage temp.

2-8°C

SMILES string

OC[C@@H](O)[C@H](O)C(O)=O.[Li+]

InChI

1S/C4H8O5.Li/c5-1-2(6)3(7)4(8)9;/h2-3,5-7H,1H2,(H,8,9);/q;+1/t2-,3+;/m1./s1

InChI key

LXIXNEPXOOEPDK-MUWMCQJSSA-N

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jorge Domínguez-Andrés et al.
Astrobiology, 20(11), 1353-1362 (2020-05-12)
The last decade has witnessed a renewed interest in space exploration. Public and private institutions are investing considerable effort toward the direct exploration of the Moon and Mars, as well as more distant bodies in the solar system. Both automated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service