Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

79252

Sigma-Aldrich

Alkyne MegaStokes dye 735

for copper catalyzed labeling

Synonym(s):

4-{2-[6-(Diethylamino)-2-benzofuranyl]ethenyl}-1-(4-pentyn-1-yl)-3-sulfo-pyridinium inner salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H26N2O4S
CAS Number:
Molecular Weight:
438.54
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

form

crystals

fluorescence

λex 586 nm; λem 735 nm in ethanol

storage temp.

−20°C

SMILES string

CCN(CC)c1ccc2cc(\C=C\c3cc[n+](CCCC#C)cc3S([O-])(=O)=O)oc2c1

InChI

1S/C24H26N2O4S/c1-4-7-8-14-25-15-13-19(24(18-25)31(27,28)29)10-12-22-16-20-9-11-21(17-23(20)30-22)26(5-2)6-3/h1,9-13,15-18H,5-8,14H2,2-3H3

InChI key

HHTYOPQTKKSRPO-UHFFFAOYSA-N

General description

The MegaStokes dyes for „click" biological labeling, licensed by LuminoChem are virtually covering the whole visible spectrum reaching the infra-red regime. Besides dyes that are being capable of participating in classical copper catalyzed 1,3-dipolar cycloaddition reaction with the counterparting function, we also provide dyes containing cyclooctyne moiety an alkyne derivative that enables copper free clicking to azides.
These dyes are noteworthy for their large Stokes-shift (120 or 125 nm). Dyes for this kind are exceptionally useful in fluorescence resonance electron transfer (FRET) applications as they do not interfere with the spectral bands of the second fluorophore a common problem in FRET technology. Azido sugars have been used for click-labeling of surface glycoproteins, recently. We have adapted this system to demonstrate the ability of our dyes to undergo bioorthogonal labeling reaction, efficiently. Prior to labeling, Chinese hamster ovary (CHO) cells were incubated with these MegaStockesdyes to test the possible cytostatic effects of these dyes. Experimental results have shown that neither dye is toxic up to 50 μM concentration.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service