Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

79182

Sigma-Aldrich

Potassium chloride 20 mM – Magnesium chloride 5 mM – TRIS Buffer pH 8.0; 50 mM – 2-Methyl-2,4-pentanediol 30% – Isopropanol 5% – HEPES Buffer pH 7.0; 50 mM – Ammonium sulfate 1.2 M solution

Synonym(s):

Crystallization solution 19/Kit-No 18839

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

form

solution

pH

7.0±0.2 ( in neat)

density

1.09 g/mL at 20 °C

storage temp.

2-8°C

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Repr. 2 - Skin Irrit. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

118.4 °F - closed cup

flash_point_c

48 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service