Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

78999

Sigma-Aldrich

Phalloidin–Atto 430LS

suitable for fluorescence

Synonym(s):

Atto 430LS-Phalloidin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

form

solid

mol wt

Mw 1359 g/mol

manufacturer/tradename

ATTO-TEC GmbH

fluorescence

λex 433 nm; λem 520.0 nm±10.0 nm in PBS, pH 7.4

suitability

suitable for fluorescence

storage temp.

−20°C

General description

Atto 430LS is a new fluorescent label featuring an extraordinary large Stokes-Shift of 114 nm. Thus its emission spectrum is almost completely separated from its absorption spectrum, making the dye highly suitable for multiplexing experiments. Atto 430LS is very hydrophilic and shows excellent water solubility. The dye exhibits a high fluorescence quantum yield, which is only slightly reduced after conjugation to biomolecules, e.g. proteins, even at high degrees of labeling (DOL).
Atto 430LS is an anionic dye. After conjugation to a substrate the dye carries a net electrical charge of -1. The fluorescence is excited most efficiently in the range 400 - 460 nm.
Phalloidin, a bicyclic heptapeptide, is a very strong binding reagent to actin. Fluorescent labeled phalloidin has become a useful tool to investigate the distribution of F-actin within the cytoskeleton of cells by fluorescence microscopy.

find more information here

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service