Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

78721

Sigma-Aldrich

Peptidoglycan from Methanobacterium sp.

Synonym(s):

PGN

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

biological source

bacterial (Methanobacterium spp.)

form

powder

storage temp.

−20°C

Application

Peptidoglycans are major components of bacterial cell walls, especially of gram-positve bacteria. Peptidoglycan are composed of alternating residues of β-(1,4) linked N-acetylglucosamine and N-acetylmuramic acid and peptide chains that create cross-linkage to create mesh structures. Peptidoglycans from different bacterial species can be compared to determine structural and compositional differences/variations and phylogenetic evolution. Peptidoglycans from different species may be used to differentiate and characterize peptidoglycan hydrolases.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Peptidoglycan structure: alternating GlcNAc and N-acetylmuramic acid cross-linked to peptides form the basic backbone.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service