Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

78674

Sigma-Aldrich

Imidazolepyruvic acid hydrobromide hydrate

≥97.0% (GC)

Synonym(s):

3-(1H-imidazol-4-yl)-2-oxopropanoic acid hydrobromide hydrate, Imidazole-4(5)-pyruvic acid hydrobromide hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6N2O3 · xHBr · yH2O
Molecular Weight:
154.12 (anhydrous free base basis)
UNSPSC Code:
12352106
PubChem Substance ID:

assay

≥97.0% (GC)

form

powder

storage temp.

2-8°C

SMILES string

OC(C(CC1=CN=CN1)=O)=O.C.C

InChI

1S/C6H6N2O3.2CH4/c9-5(6(10)11)1-4-2-7-3-8-4;;/h2-3H,1H2,(H,7,8)(H,10,11);2*1H4

InChI key

SHQBBHAXDQVOGW-UHFFFAOYSA-N

Biochem/physiol Actions

Histidinemia is one of the most frequent of the inborn errors of metabolism and is easily diagnosed on the basis of increased excretion of imidazolepyruvic acid in the urine.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tao Zhang et al.
Journal of proteome research, 12(1), 505-512 (2012-11-21)
To evaluate the application of urinary metabolomics on discovering potential biomarkers for epithelial ovarian cancer (EOC), urine samples from 40 preoperative EOC patients, 62 benign ovarian tumor (BOT) patients, and 54 healthy controls were collected and analyzed with ultraperformance liquid
THE METABOLISM OF HISTIDINE. VI. HISTIDINEMIA AND IMIDAZOLEPYRUVIC ACIDURIA.
R C BALDRIDGE et al.
The Journal of biological chemistry, 239, 1557-1559 (1964-05-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service