Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

78581

Sigma-Aldrich

Fluorescent red NIR 730

suitable for fluorescence

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H48N2O6S
Molecular Weight:
660.86
MDL number:
UNSPSC Code:
12352108
NACRES:
NA.32

form

powder

solubility

DMSO: soluble

fluorescence

λex 680 nm; λem ~755 nm in 0.1 M phosphate pH 7.0

suitability

suitable for fluorescence

storage temp.

2-8°C

General description

Fluorescent red 730 is a fluorescent dye for the near infrared area. Extremely long wavelength fluorescence is especially well suited for applications where sample autofluorescence might be critical or penetration of tissue or other matrix is required. Fluorescent red 730 shows strong fluorescence (molar absorption 185 000 M-1cm-1)

Analysis Note

further Exc.: 730 nm (results in a lower emission)

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service